Merck
Anxiolytic-like activity of SB-205384 in the elevated plus-maze test in mice.
Psicothema (2007-02-14)
José Francisco Navarro, Estrella Burón, Mercedes Martín-López
ZUSAMMENFASSUNG
Recent studies point to a major role for alpha2-containing GABA-A receptors in modulating anxiety. However, the possible implication of GABA-A receptors containing the alpha3 subunit on anxiety is less known. The aim of this study was to examine the effects of SB-205384 (0.5-4 mg/kg, i.p.), an alpha3 subunit positive modulator of GABA-A receptor, on anxiety tested in the elevated plus-maze in male mice, using classical and ethological parameters. Mice treated with SB-205384 showed an increase in the frequency of entries and the time spent in open arms, as well as a reduction in the time spent in closed arms, as compared with the control group. A notable increase of "head-dipping" unprotected and a reduction of "stretched-attend posture" protected was also evident. These findings indicate that SB-205384 exhibits an anxiolytic-like profile in the elevated plus-maze test, suggesting that GABA-A receptors which contain the alpha3 subunit might be involved in regulation of anxiety.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SB 205384, solid