Merck
  • Resynchronization of estrus in beef cattle: ovarian function, estrus and fertility following progestin treatment and treatments to synchronize ovarian follicular development and estrus.
Resynchronization of estrus in beef cattle: ovarian function, estrus and fertility following progestin treatment and treatments to synchronize ovarian follicular development and estrus.
The Canadian veterinary journal = La revue veterinaire canadienne (2007-02-22)
Marcos G Colazo, John P Kastelic, Julie A Small, Randy E Wilde, Douglas R Ward, Reuben J Mapletoft
ZUSAMMENFASSUNG
The objective was to optimize rebreeding of nonpregnant, previously inseminated beef cattle. In Experiment 1, 43 cows received a used intravaginal progesterone-releasing insert (IVPRI; Days 0-7) 12.3 d after ovulation and received concurrently no treatment, 100 microg gonadotropin releasing hormone (GnRH), 1 mg estradiol cypionate (ECP), or 150 mg progesterone. Emergence of a new ovarian follicular wave was most synchronous (P < 0.0001) in the GnRH group. In Experiment 2, 675 heifers were given GnRH or no treatment on Day 0, fed melengestrol acetate (MGA; 0.5 mg/head/d) from Days 0-5 (Day 0 = 13-14 d after timed insemination; TAI), given 0.5 mg ECP or nothing on Day 7, and reinseminated 6-12 h after onset of estrus. Estrus was more synchronous (P < 0.05) in heifers given GnRH versus no treatment on Day 0. In Experiment 3, 317 TAI heifers were resynchronized with either MGA or a used IVPRI with or without ECP on Day 7; estrus was more synchronous (P < 0.05) and pregnancy rates were higher (54.1% versus 39.2%, P < 0.05) in heifers given a used IVPRI than those fed MGA. For resynchronization of heifers, pregnancy rates were not significantly improved with GnRH treatment, but were higher with a used IVPRI than with MGA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Estradiol-17-Cypionat