Merck
  • The alpha-methylene-gamma-butyrolactone moiety in dehydrocostus lactone is responsible for cytoprotective heme oxygenase-1 expression through activation of the nuclear factor E2-related factor 2 in HepG2 cells.
The alpha-methylene-gamma-butyrolactone moiety in dehydrocostus lactone is responsible for cytoprotective heme oxygenase-1 expression through activation of the nuclear factor E2-related factor 2 in HepG2 cells.
European journal of pharmacology (2007-03-30)
Gil-Saeng Jeong, Hyun-Ock Pae, Sun-Oh Jeong, Youn-Chul Kim, Tae-Oh Kwon, Ho Sub Lee, Nam-Song Kim, Seok Don Park, Hun-Taeg Chung
ZUSAMMENFASSUNG
Inducible heme oxygenase (HO)-1 acts against oxidants that are thought to play a major role in the pathogenesis of several diseases. The alpha-methylene-gamma-butyrolactone (CH2-BL) structural unit, which characterizes a group of naturally occurring sesquiterpene lactones, is known to possess numerous biological activities. In the present study, we evaluated dehydrocostus lactone possessing CH2-BL moiety, one of the bioactive constituents of the medicinal plant Saussurea lappa, as an inducer of cytoprotective HO-1. In HepG2 cells, treatment with dehydrocostus lactone induced HO-1 expression and increased HO activity in a concentration-dependent manner. Similar results were also observed when the cells were incubated with CH2-BL, a parent structure of dehydrocostus lactone. In contrast, mokko lactone, a reduced product of dehydrocostus lactone, and alpha-methyl-gamma-butyrolactone (CH3-BL), a parent structure of mokko lactone, did not induce HO-1 expression. Pretreatment with either dehydrocostus lactone or CH2-BL for 6 h protected the cells from hydrogen peroxide-mediated toxicity, whereas mokko lactone or CH3-BL failed to exert a cytoprotective action. Inhibition of HO-1 expression by HO-1 small interfering RNA (siRNA) abrogated cellular protection afforded by dehydrocostus lactone or CH2-BL. In addition, dehydrocostus lactone caused the nuclear accumulation of the nuclear factor E2-related factor 2 (Nrf2) and increased the promoter activity of antioxidant response element (ARE). Using Nrf2 siRNA, Nrf2 activation was confirmed to contribute to cytoprotective HO-1 expression by dehydrocostus lactone or CH2-BL. Collectively, our findings suggest that CH2-BL moiety in dehydrocostus lactone increases cellular resistance to oxidant injury in HepG2 cells, presumably through Nrf2/ARE-dependent HO-1 expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Methylen-γ-butyrolacton, 97%