Merck
  • Determination of plant resistance to carbamate herbicidal compounds inhibiting cell division and early growth by seed and plantlets bioassays.
Determination of plant resistance to carbamate herbicidal compounds inhibiting cell division and early growth by seed and plantlets bioassays.
Nature protocols (2007-04-05)
Yael Weiss, Baruch Rubin, Avidor Shulman, Irina Ben Shir, Ehud Keinan, Shmuel Wolf
ZUSAMMENFASSUNG
Herbicide-resistant plants can be generated by either traditional breeding procedures or genetic engineering. Analyses of plant responses to a newly developed herbicide or the tolerance level of a newly developed plant line to a given herbicide are based on various bioassays. Here, we describe several methods for quantitative measurements of plants' responses to propham application, as a model herbicide of the carbamate family. Dose-response assays include seed germination and analyses of shoot and root elongation on paper. To better reflect the natural interaction between the plant, the soil and the herbicide, a protocol for germination and root elongation on sand is described. Finally, a more sensitive bioassay is based on plant growth on agar medium. The described protocols are simple, reproducible and can be easily adopted for a variety of plant species and for various herbicides. Plants' response to a given herbicide can be determined within a few weeks.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Propham, PESTANAL®, analytical standard