- Function-oriented synthesis: studies aimed at the synthesis and mode of action of 1alpha-alkyldaphnane analogues.
Function-oriented synthesis: studies aimed at the synthesis and mode of action of 1alpha-alkyldaphnane analogues.
Organic letters (2007-04-06)
Paul A Wender, Noel D'Angelo, Vassil I Elitzin, Martin Ernst, Eileen E Jackson-Ugueto, John A Kowalski, Sharon McKendry, Markus Rehfeuter, Robert Sun, David Voigtlaender
PMID17408281
ZUSAMMENFASSUNG
[reaction: see text] An efficient synthetic route to the ABC tricyclic core of 1alpha-alkyldaphnanes has been developed. The conformational bias imparted by the C6-C9 oxo-bridge of BC-ring system 12 was used to elaborate the ABC-ring system precursor including the introduction of the beta-C5 hydroxyl group. A completely diastereoselective palladium-catalyzed enyne cyclization was then employed to establish the A-ring with a C1 appendage.