Merck
  • A cryptic PKS-NRPS gene locus in the plant commensal Pseudomonas fluorescens Pf-5 codes for the biosynthesis of an antimitotic rhizoxin complex.
A cryptic PKS-NRPS gene locus in the plant commensal Pseudomonas fluorescens Pf-5 codes for the biosynthesis of an antimitotic rhizoxin complex.
Organic & biomolecular chemistry (2007-07-05)
Nicole Brendel, Laila P Partida-Martinez, Kirstin Scherlach, Christian Hertweck
ZUSAMMENFASSUNG
Targeted gene inactivation and metabolic profiling revealed that the cryptic PKS-NRPS gene cluster in the genome of the plant commensal Pseudomonas fluorescens Pf-5 codes for the biosynthesis of antiproliferative and antifungal rhizoxin derivatives.