Merck
  • Nonenzymatic oxidative steps accompanying action of the cytochrome P450 enzymes StaP and RebP in the biosynthesis of staurosporine and rebeccamycin.
Nonenzymatic oxidative steps accompanying action of the cytochrome P450 enzymes StaP and RebP in the biosynthesis of staurosporine and rebeccamycin.
Journal of the American Chemical Society (2007-08-21)
Annaleise R Howard-Jones, Christopher T Walsh