Merck
  • Comment on "Midodrine hydrochloride and L-threo-3,4-dihydroxy-phenylserine preserve cerebral blood flow in hemodialysis patients with orthostatic hypotension" by Fujisaki et al.
Comment on "Midodrine hydrochloride and L-threo-3,4-dihydroxy-phenylserine preserve cerebral blood flow in hemodialysis patients with orthostatic hypotension" by Fujisaki et al.
Therapeutic apheresis and dialysis : official peer-reviewed journal of the International Society for Apheresis, the Japanese Society for Apheresis, the Japanese Society for Dialysis Therapy (2007-09-12)
Vijay Sharma, Bernard P Chan
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-DOPS, ≥98% (HPLC)