Merck
  • Complexation of tauro- and glyco-conjugated bile salts with three neutral beta-CDs studied by ACE.
Complexation of tauro- and glyco-conjugated bile salts with three neutral beta-CDs studied by ACE.
Electrophoresis (2007-09-26)
René Holm, Henrik V Nicolajsen, Rune A Hartvig, Peter Westh, Jesper Ostergaard
ZUSAMMENFASSUNG
Complexation of the bile salts (BS) taurocholate, tauro-beta-muricholate, taurodeoxycholate, taurochenodeoxycholate, glycocholate, glycodeoxycholate, and glycochenodeoxycholate common in rat, dog, and man with natural beta-CD and the chemically modified beta-CDs 2-hydroxypropyl-beta-CD and 2-O-methyl-beta-CD was studied using mobility shift ACE. The CDs were selected due to their frequent use in preformulation and drug formulation as oral excipients for the solubilization of drug substances with low aqueous solubility. ACE was demonstrated to be a feasible and efficient technique for investigation of the interactions between BS and beta-CDs. All the investigated BS possessed affinity for the three CDs with stability constants ranging from 2x10(3) to 4x10(5) M(-) (1). The requirements and assumptions related to the use of ACE for estimating high affinity stability constants were discussed. The extent and pattern of hydroxylation significantly influenced the affinity of the glyco- and tauro-conjugated BS toward the beta-CDs (chenodeoxycholates > deoxycholates > cholates) whereas the nature of the beta-CD derivatization and BS conjugation played a minor role only. The results indicate that displacement of drug substances from beta-CD inclusion complexes is likely to occur in the small intestine where BS are present potentially influencing drug bioavailability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sodium taurolithocholate