Merck
Effects of octreotide in acute hemorrhagic necrotizing pancreatitis in rats.
Journal of gastroenterology and hepatology (2007-10-05)
Frank Axel Wenger, Maik Kilian, Ina Heukamp, Thomas Foitzik, Christoph Andreas Jacobi, Hans Guski, Ingolf Schimke, Joachim Michael Müller
ZUSAMMENFASSUNG
Octreotide is considered to reduce exocrine pancreatic secretion in acute hemorrhagic necrotizing pancreatitis decreasing pancreatic autodigestion. The aim of this study was to determine whether octreotide also has antioxidative effects in acute pancreatitis. Additionally time and dose of application were of interest. Ninety male Sprague-Dawley rats were randomized into six groups (n = 15). Group 1 underwent a laparotomy, and animals in groups 2-6 received intraductal glycodeoxycholic acid followed by intravenous cerulein. Groups 3 and 4 were injected with 0.5 mg octreotide, while groups 5 and 6 received continuous intravenous infusion of 0.05 mg octreotide/h for 10 h. Treatment was initiated 6 hours after induction of pancreatitis (IP) in groups 3 and 5, and 14 h after IP in groups 4 and 6. At 24 h after IP all animals were killed and each pancreas was analyzed histopathologically. In addition, levels of pancreatic lipid peroxidation protective enzymes glutathione-peroxidase (GSH-Px) and superoxide dismutase (SOD) as well as lipid peroxidation via thiobarbituric acid reactive substances (TBARS) were determined. Early bolus application of octreotide reduced severity of histopathological changes in acute pancreatitis and decreased lipid peroxidation in pancreatic tissue samples; however, late bolus application and continuous intravenous infusion did not influence pancreatitis or lipid peroxidation. Octreotide seems to have a dose- and time-dependent effect on histopathology and lipid peroxidation in a model of pancreatitis in rats.