Merck
Production of D-phenylglycine-related amino acids by immobilized microbial cells.
Biochimie (1980-01-01)
H Yamada, S Shimizu, H Shimada, Y Tani, S Takahashi, T Ohashi
ZUSAMMENFASSUNG
Bacterial dihydropyrimidinase was shown to catalyze the hydrolytic cleavage of various 5-substituted hydantoins to the corresponding N-carbamyl-D-amino acids under alkaline conditions. Therefore, an enzymatic method for preparing the D-forms of phenylglycine-related amino acids was developed using immobilized bacterial cells with high enzyme activity. Alkalophilic bacteria were a good enzyme source for this process. The process is simple and economical for use in the production of various amino acids with the D-configuration.