- Synthesis and asymmetric mono-cyclopalladation of 1,1'-di(alpha-dimethylamino)ethylferrocene.
Synthesis and asymmetric mono-cyclopalladation of 1,1'-di(alpha-dimethylamino)ethylferrocene.
Molecules (Basel, Switzerland) (2007-11-17)
Viatcheslav I Sokolov, Ludmila L Troitskaya, Veronica L Bondareva-Don
PMID18007334
ZUSAMMENFASSUNG
Two-step synthesis of 1,1'-di(alpha-dimethylamino)ethylferrocene is described in details. Cyclopalladation of this diamine involving one or two amino groups is reported and the product of asymmetric mono-cyclopalladation is fully characterized.