Merck
Piezoelectric properties of (K,Na)NbO3 thin films deposited on (001)SrRuO3/Pt/MgO substrates.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control (2008-02-16)
Isaku Kanno, Takuya Mino, Shuichiro Kuwajima, Takaaki Suzuki, Hidetoshi Kotera, Kiyotaka Wasa
ZUSAMMENFASSUNG
(K(x),Na(1-x))NbO(3) (KNN) thin films were deposited on (001)SrRuO(3)/(001)Pt/(001)MgO substrates by RF-magnetron sputtering, and their piezoelectric properties were investigated. The x-ray diffraction measurements indicated that the KNN thin films were epitaxially grown with the c-axis orientation in the perovskite tetragonal system. The lattice constant of the c-axis increased with increasing concentrations of potassium. The KNN thin films showed typical ferroelectric behavior; the relative dielectric constant epsilon(r) was 270 to approximately 320. The piezoelectric properties were measured from the tip displacement of the KNN/MgO unimorph cantilevers; the transverse piezoelectric coefficient epsilon*(31) (= d(31)/s(E)(11)) of KNN (x = 0) thin films was calculated to be -0.9 C/m(2). On the other hand, doping of potassium caused an increase in the piezoelectric properties, and the KNN (x = 0.16) films showed a relatively large transverse piezoelectricity of epsilon*(31) = -2.4 C/m(2).