Merck
  • Isobolographic analysis of epinephrine with bupivacaine, dextromethorphan, 3-methoxymorphinan, or dextrorphan on infiltrative anesthesia in rats: dose-response studies.
Isobolographic analysis of epinephrine with bupivacaine, dextromethorphan, 3-methoxymorphinan, or dextrorphan on infiltrative anesthesia in rats: dose-response studies.
Regional anesthesia and pain medicine (2008-02-27)
Yu-Wen Chen, Kuo-Sheng Liu, Jhi-Joung Wang, Willy Chou, Ching-Hsia Hung
ZUSAMMENFASSUNG
The aims of this study were to establish the potencies of epinephrine, bupivacaine, dextromethorphan, 3-methoxymorphinan, and dextrorphan and evaluate interactions of epinephrine with bupivacaine, dextromethorphan, 3-methoxymorphinan, or dextrorphan as an infiltrative anesthetic. Bupivacaine, a common and long-acting local anesthetic, was used as control. Dose-dependent responses of epinephrine, dextromethorphan, 3-methoxymorphinan, and dextrorphan on cutaneous analgesia were compared with bupivacaine in rats. The interactions of drugs were evaluated via an isobolographic analysis. We found that epinephrine, bupivacaine, dextromethorphan, 3-methoxymorphinan, and dextrorphan produced a dose-dependent local anesthetic effect as infiltrative cutaneous analgesia. Relative potencies were epinephrine > bupivacaine > dextromethorphan > 3-methoxymorphinan > dextrorphan (P < .01 for each comparison). Coadministration of bupivacaine with epinephrine produced a synergistic effect, and coadministration of dextromethorphan, 3-methoxymorphinan, or dextrorphan with epinephrine produced an additive effect. Epinephrine, dextromethorphan, 3-methoxymorphinan, and dextrorphan are known to have local anesthetic effects as infiltrative cutaneous analgesia in rats. Epinephrine increased the potency of bupivacaine, but not dextromethorphan, 3-methoxymorphinan, or dextrorphan as an infiltrative anesthetic. The cutaneous analgesic effects of adding epinephrine to dextromethorphan, 3-methoxymorphinan, or dextrorphan, are similar to combinations of 2 local anesthetics.