Merck
  • A convenient preparation of a disaccharide motif and its role in the cytotoxicity of the triterpenoid saponin, alpha-hederin.
A convenient preparation of a disaccharide motif and its role in the cytotoxicity of the triterpenoid saponin, alpha-hederin.
Archives of pharmacal research (2008-05-16)
Seong-Cheol Bang, Hyun-Hee Seo, Hye-Rim Shin, Ki-Cheul Lee, Le Tuan Anh Hoang, Sang-Hun Jung
ZUSAMMENFASSUNG
The sugar structures of triterpenoid saponins, such as alpha-hederin, are intimately associated with their antitumor activities and other biological activities. The alpha-L: -rhamnopyranosyl-(1-->2)-alpha-L: -arabinopyranoside group of alpha-hederin alters the cytotoxicity of its aglycon, hederagenin. This study explored the role of this saccharide unit in the cytotoxic effect of alpha-hederin and the possibility of its use as a carrier moiety in prodrugs of anticancer agents. A new convenient and practical procedure for the preparation of 4-methoxybenzoyl-2,3,4-tri-O-benzoyl-alpha-L: -rhamnopyranosyl-(1-->2)-3,4-O-dibenzoyl-beta-L: -arabinopyranoside (2) from 4-methoxybenzoyl-beta-L: -arabinopyranoside was accomplished using four steps with an overall yield of 63%. The use of BF(3)-OEt(2) as a catalyst in the glycosylation step in this procedure had a large advantage over the TMSOTf catalyst used in the usual method. Moreover, the key intermediate obtained in this procedure, 4-methoxybenzoyl-2,3,4-tri-O-benzoyl-alpha-L: -rhamnopyranosyl-(1-->2)-alpha-L: -arabinopyranoside (7), was selectively transformed to 4-methoxybenzoyl-2,3,4-tri-O-benzoyl-alpha-L: -rhamnopyranosyl-(1-->2)-4-O-acetyl-alpha-L: -arabinopyranoside (9) and 4-methoxybenzoyl-2,3,4-tri-O-benzoyl-alpha-L: -rhamnopyranosyl-(1-->2)-3-O-benzoyl-beta-L: -arabinopyranoside (10). These derivatives did not show any cytotoxicity against human cancer cell lines. Thus the 3-O-alpha-L: -rhamnopyranosyl-(1-->2)-alpha-L: -arabinopyranoside could be used as a nontoxic carrier moiety to enhance the activity of anticancer drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
α-Hederin, primary reference standard