- Nanoparticle-loaded magnetophoretic vesicles.
Nanoparticle-loaded magnetophoretic vesicles.
Journal of the American Chemical Society (2008-05-20)
Maren Krack, Heinrich Hohenberg, Andreas Kornowski, Peter Lindner, Horst Weller, Stephan Förster
PMID18484723
ZUSAMMENFASSUNG
Magnetic nanoparticles have been assembled into the bilayer membrane of block copolymer vesicles. The nanoparticles decorate the hydrophobic/hydrophilic interface, which leads to bridging of adjacent bilayers and the formation of oligo-lamellar vesicles. The nanoparticle uptake of the vesicles is sufficiently high to become magnetophoretic in external magnetic fields as shown by video microscopy.
MATERIALIEN