Merck
  • The hemodynamic effects of rebaudioside A in healthy adults with normal and low-normal blood pressure.
The hemodynamic effects of rebaudioside A in healthy adults with normal and low-normal blood pressure.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-06-17)
K C Maki, L L Curry, M C Carakostas, S M Tarka, M S Reeves, M V Farmer, J M McKenney, P D Toth, S L Schwartz, B C Lubin, M R Dicklin, A C Boileau, J D Bisognano
ZUSAMMENFASSUNG
Rebaudioside A and stevioside are steviol glycosides extracted from the plant Stevia rebaudiana Bertoni and are used in several countries as food and beverage sweeteners. This randomized, double-blind trial evaluated the hemodynamic effects of 4weeks consumption of 1000mg/day rebaudioside A vs. placebo in 100 individuals with normal and low-normal systolic blood pressure (SBP) and diastolic blood pressure (DBP). Subjects were predominantly female (76%, rebaudioside A and 82%, placebo) with a mean age of approximately 41 (range 18-73) years. At baseline, mean resting, seated SBP/DBP was 110.0/70.3mmHg and 110.7/71.2mmHg for the rebaudioside A and placebo groups, respectively. Compared with placebo, rebaudioside A did not significantly alter resting, seated SBP, DBP, mean arterial pressure (MAP), heart rate (HR) or 24-h ambulatory blood pressures responses. These results indicate that consumption of as much as 1000mg/day of rebaudioside A produced no clinically important changes in blood pressure in healthy adults with normal and low-normal blood pressure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rebaudiosid A, ≥96% (HPLC)