Merck
  • Dopamine acting through D2 receptors modulates the expression of PSA-NCAM, a molecule related to neuronal structural plasticity, in the medial prefrontal cortex of adult rats.
Dopamine acting through D2 receptors modulates the expression of PSA-NCAM, a molecule related to neuronal structural plasticity, in the medial prefrontal cortex of adult rats.
Experimental neurology (2008-08-23)
Esther Castillo-Gómez, María Angeles Gómez-Climent, Emilio Varea, Ramón Guirado, José Miguel Blasco-Ibáñez, Carlos Crespo, Francisco José Martínez-Guijarro, Juan Nácher
ZUSAMMENFASSUNG
A "neuroplastic" hypothesis proposes that changes in neuronal structural plasticity may underlie the aetiology of depression and the action of antidepressants. The medial prefrontal cortex (mPFC) is affected by this disorder and shows an intense expression of the polysialylated form of the neural cell adhesion molecule (PSA-NCAM), a plasticity-associated molecule, which is expressed mainly in interneurons. The monoamines serotonin, dopamine and noradrenaline are the principal targets of antidepressant action. Pharmacological manipulation of serotonin levels regulates synaptophysin and PSA-NCAM expression in the adult mPFC. However, the involvement of structural plasticity on the antidepressant effects of dopamine has not been well explored yet. Using immunohistochemistry, we have studied the relationship between dopaminergic fibers and PSA-NCAM expressing neurons in the mPFC and the expression of D2 receptors. In order to evaluate the effects of dopamine in neuronal structural plasticity and on inhibitory neurotransmission, we have analyzed the expression of synaptophysin, PSA-NCAM and GAD67 in the mPFC after cortical dopamine depletion with 6-OHDA and after chronic treatments with the D2 receptor antagonist haloperidol or the D2 receptor agonist PPHT. Many dopaminergic fibers were observed in close apposition to PSA-NCAM expressing neurons and 76% of these cells co-expressed D2 receptor. Both haloperidol treatment and 6-OHDA injection reduced significantly PSA-NCAM, synaptophysin and GAD67 expression in the mPFC. Conversely, PPHT treatment increased the expression of these molecules. Our results give support to the "neuroplastic" hypothesis of depression, suggesting that dopamine acting on D2 receptors may modulate neuronal structural plasticity and inhibitory neurotransmission through changes in PSA-NCAM expression.