Merck
  • Comparative cytotoxic effects of acetaminophen (N-acetyl-p-aminophenol), a non-hepatotoxic regioisomer acetyl-m-aminophenol and their postulated reactive hydroquinone and quinone metabolites in monolayer cultures of mouse hepatocytes.
Comparative cytotoxic effects of acetaminophen (N-acetyl-p-aminophenol), a non-hepatotoxic regioisomer acetyl-m-aminophenol and their postulated reactive hydroquinone and quinone metabolites in monolayer cultures of mouse hepatocytes.
Biochemical pharmacology (1991-08-08)
J A Holme, J K Hongslo, C Bjørge, S D Nelson
ZUSAMMENFASSUNG
Toxic effects of acetaminophen (paracetamol, N-acetyl-p-aminophenol, APAP) in monolayer cultures of mouse hepatocytes developed over a period of 18 hr. N-Acetyl-m-aminophenol (AMAP) was approximately 10-fold less toxic than APAP, despite the fact that it bound covalently to a greater extent to hepatocyte macromolecules. AMAP did not deplete glutathione to as great an extent as APAP, indicating that their reactive metabolites may bind to different proteins or that oxidative damage in addition to arylation of proteins may be involved in the development of cell death. The toxicity of 3-methoxy-acetyl-p-aminophenol was similar to that of APAP, whereas the other hydroquinone and quinone metabolites were 8-10 times more cytotoxic than APAP. The potencies of these analogs were in the order: acetyl-m-aminophenol-p-benzoquinoneimine greater than or equal to 2,5-dihydroxyacetanilide greater than or equal to 3-methoxy-p-benzoquinone greater than or equal to N-acetyl-p-benzoquinone imine (NAPQI) greater than or equal to acetyl-m-aminophenol-o-benzoquinone greater than or equal to 3-hydroxy-acetyl-p-aminophenol. The relative toxic potencies of the hydroquinone and quinone metabolites of AMAP were comparable to that of NAPQI, and do not readily explain the marked difference between the cytotoxic effects of AMAP and APAP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Acetamidphenol, 97%