Merck
  • Acaricidal activity of limonene, limonene oxide and beta-amino alcohol derivatives on Rhipicephalus (Boophilus) microplus.
Acaricidal activity of limonene, limonene oxide and beta-amino alcohol derivatives on Rhipicephalus (Boophilus) microplus.
Veterinary parasitology (2008-08-30)
Stela Regina Ferrarini, Mônica Oliveira Duarte, Ricardo G da Rosa, Verônica Rolim, Vera Lucia Eifler-Lima, Gilsane von Poser, Vera Lucia Sardá Ribeiro
ZUSAMMENFASSUNG
Limonene, limonene oxide and eight beta-amino alcohol derivatives obtained by synthesis were investigated for the effect on egg hatchability and mortality rates of newly hatched larvae of the cattle tick Rhipicephalus (Boophilus) microplus. At the doses between 10 microg/ml and 2.5 microg/ml all the compounds were highly lethal to the larvae and some of them showed activity at lower concentrations. The effect on the eggs hatchability was observed in all the treatments.