Merck
  • Lack of tolerance and morphine-induced cross-tolerance to the analgesia of chimeric peptide of Met-enkephalin and FMRFa.
Lack of tolerance and morphine-induced cross-tolerance to the analgesia of chimeric peptide of Met-enkephalin and FMRFa.
Peptides (2008-10-22)
Kshitij Gupta, Ishwar Dutt Vats, Yogendra Kumar Gupta, Kishwar Saleem, Santosh Pasha
ZUSAMMENFASSUNG
Chimeric peptide of Met-enkephalin and FMRFa (YGGFMKKKFMRFa-YFa), a kappa-opioid receptor specific peptide, did not induce tolerance and cross-tolerance effects to its analgesic action on day 5 after pretreatment with either YFa or morphine for 4 days. However, pretreatment with YFa for 4 days led to the development of cross-tolerance to the analgesic effects of morphine and also 4 days of pretreatment of morphine resulted in the expression of tolerance to its own analgesic effects. Similar expression of tolerance and cross-tolerance were also observed when YFa was compared with the kappa receptor agonist peptide dynorphin A(1-13) [DynA(1-13)]. Cross-tolerance effects between YFa and DynA(1-13) analgesia were also not observed on day 5. Interestingly, when YFa and DynA(1-13) were tested for their analgesic effects for 5 days, reduction in analgesia on day 3 was observed in case of DynA(1-13) whereas YFa maintained its analgesia for 5 days. Thus, chimeric peptide YFa may serve as a useful probe to understand pain modulation and expression of tolerance and cross-tolerance behavior with other opioids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dynorphin A Porcine Fragment 1-13, ≥97% (HPLC)