Merck
Biological activities of novel derivatives of DIF-1 isolated from Dictyostelium.
Biochemical and biophysical research communications (2008-11-04)
Haruhisa Kikuchi, Shinya Ishiko, Yoshiteru Oshima, Naomi Gokan, Kohei Hosaka, Yuzuru Kubohara
ZUSAMMENFASSUNG
The differentiation-inducing factor-1 (DIF-1) is a lipophilic signal molecule (chlorinated alkylphenone) that induces stalk cell differentiation in the cellular slime mold Dictyostelium discoideum. In addition, DIF-1 and its derivatives have been shown to possess anti-leukemic activity and glucose consumption-promoting activity in vitro in mammalian cells. In this study, to assess the chemical structure-effect relationship of DIF-1, we synthesized eight derivatives of DIF-1 and investigated their stalk cell-inducing activity in Dictyostelium cells and pharmacological activities in mammalian cells. Of the derivatives, two amide derivatives of DIF-1, whose hydrophobic indexes are close to that of DIF-1, induced stalk cell differentiation as strongly as DIF-1 in Dictyostelium cells. It was also found that some derivatives suppressed cell growth in human K562 leukemia cells and promoted glucose consumption in mouse 3T3-L1 cells. These results give us valuable information as to the chemical structure-effect relationship of DIF-1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DIF-1, ≥98% (HPLC)