- Acaricidal effects of the essential oil of Origanum minutiflorum (Lamiaceae) against Rhipicephalus turanicus (Acari: Ixodidae).
Acaricidal effects of the essential oil of Origanum minutiflorum (Lamiaceae) against Rhipicephalus turanicus (Acari: Ixodidae).
Veterinary parasitology (2008-12-19)
Huseyin Cetin, James E Cilek, Levent Aydin, Atila Yanikoglu
PMID19091479
ZUSAMMENFASSUNG
The acaricidal effects of the volatile essential oil Origanum minutiflorum O. Schwarz & P.H. Davis (Lamiaceae) against adult Rhipicephalus turanicus was evaluated at a variety of concentrations and exposure times. Generally tick mortality increased with concentration and exposure. Ticks exposed to vapors from cotton wicks containing at least 10mul/L resulted in complete (100%) mortality at 120min. The major constituent of essential oil obtained from the plant material of O. minutiflorum was carvacrol.