Merck
  • Phosphatase inhibition augments anti-CD22-mediated signaling and cytotoxicity in non-hodgkin's lymphoma cells.
Phosphatase inhibition augments anti-CD22-mediated signaling and cytotoxicity in non-hodgkin's lymphoma cells.
Leukemia research (2009-02-25)
Robert T O'Donnell, David Pearson, Hayes C McKnight, Ya Peng Ma, Joseph M Tuscano
ZUSAMMENFASSUNG
CD22 is a cell-surface molecule found on most B-cell lymphomas (NHL). HB22.7 is an anti-CD22 antibody that blocks CD22 ligand binding, initiates signaling, and kills NHL cells. The SHP-1 tyrosine phosphatase is disproportionately associated with the cytoplasmic domain of CD22. Sodium orthovanadate (NaV) and dephostatin (DP) are phosphatase inhibitors. The interaction of SHP-1 with CD22 presents an opportunity to manipulate CD22-mediated signaling effects. NaV caused dose dependent killing of NHL cells in vitro; when HB22.7 was given with NaV, antibody-mediated cell death increased. NaV caused a substantial increase in CD22-mediated SAPK and ERK-1/2 activation when CD22 was crosslinked by HB22.7; NaV did not significantly affect IgM-mediated signals. Studies using Raji NHL cells stably transfected with a SHP-1 dominant negative (DN) confirmed that these observations were due to SHP-1 inhibition. The relatively specific association of SHP-1 with CD22 suggests that CD22-specific signaling may be altered by phosphatase inhibition in ways that could prove useful for anti-CD22-based immunotherapy.