Merck
  • Association between efficacy and body weight or body mass index for two low-dose oral contraceptives.
Association between efficacy and body weight or body mass index for two low-dose oral contraceptives.
Contraception (2009-05-16)
Ronald T Burkman, Alan C Fisher, George J Wan, Christopher E Barnowski, Katherine D LaGuardia
ZUSAMMENFASSUNG
This analysis investigated the association of oral contraceptive efficacy with body weight and body mass index (BMI) for hypothesis-generating purposes. Data were from a randomized, parallel-group trial of 180/215/250 mcg of norgestimate (NGM)/25 mcg of ethinyl estradiol (EE) (given to 1671 women) and 1 mg of norethindrone acetate (NETA)/20 mcg of EE (given to 1139 women). Pregnancies were evaluated across BMI deciles and by BMI and body weight dichotomies. A Pearl index was calculated for each treatment group. The relative risk (RR) of pregnancy was calculated with a Cox proportional hazards model. The Pearl index for women who received NGM/EE was 2.36 [95% confidence interval (CI)=1.33-3.40]; for those who received NETA/EE, the Pearl index was 3.29 (95% CI=1.81-4.77). Consistent, weak positive associations between weight and pregnancy risk were found. Overall, for women with a BMI >or=25 kg/m(2) (compared with women with a BMI <25 kg/m(2)), the RR of pregnancy was 1.84 (95% CI=0.98-3.45); that for women who received NGM/EE was 1.39 (95% CI=0.57-3.40), whereas that for women who received NETA/EE was 2.49 (95% CI=1.01-6.13). For women with a body weight >or=70 kg (compared with women with a body weight <70 kg), the RR was 1.25 (95% CI=0.63-2.46); that for women who received NGM/EE was 1.41 (95% CI=0.56-3.54), whereas that for women who received NETA/EE was 1.12 (95% CI=0.40-3.12). Women in the higher body weight or BMI category showed a small increase in the risk of pregnancy with these oral contraceptives, but this increase was not statistically significant overall or for either formulation studied.