Merck
  • Synthesis of tetrahydro-beta-carbolines and tetrahydroisoquinolines fused to pyrrolidines and solution-phase parallel acylation.
Synthesis of tetrahydro-beta-carbolines and tetrahydroisoquinolines fused to pyrrolidines and solution-phase parallel acylation.
Journal of combinatorial chemistry (2009-05-29)
Daniel Blanco-Ania, Pedro H H Hermkens, Leo A J M Sliedregt, Hans W Scheeren, Floris P J T Rutjes
ZUSAMMENFASSUNG
A structurally diverse library of potentially pharmacologically important compounds employing classical synthesis methods is described. These compounds are synthesized from beta-aryl pyrrolidines, providing products with the 2-arylethyl amine moiety, a structural feature often encountered in compounds active in the central nervous system. Tri- and tetracyclic scaffolds were obtained using the Pictet-Spengler reaction, resulting in hexahydropyrrolo[3,4-c]isoquinolines 1-3, an octahydropyrrolo[3',4':5,6]pyrido[3,4-b]indole 4, and a hexahydrofuro[2,3-d]pyrrolo[3,4-b]pyridine 5. These scaffolds were further derivatized in parallel fashion to make a 32-membered amide library with yields from 62 to 100% (90% average) and purities from 63 to 100% (93% average).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indol, 98%