- [The central nervous system and inflammation: the role of opioid peptides].
[The central nervous system and inflammation: the role of opioid peptides].
Medicina (Florence, Italy) (1990-01-01)
A Mazzone, G Ricevuti, A Fioravanti, D Pasotti, A R Vitto, L Mirto, C Vignali, F Avato, A Notario
PMID1974317
ZUSAMMENFASSUNG
Inhibition of human granulocyte aggregation was observed in the presence of morphine in a naloxone stereoselective way, while the opioid peptides were ineffective. In comparative studies Ca(+)+ (A23187) and dynorphin 1-9, per se, induced stimulation of arachidonic acid metabolites from granulocytes. In this regard dynorphin 1-9 may function as a mediator between the central nervous system and the inflammation.