Merck
Wheat germ oil in larval diet influences gene expression in adult oriental fruit fly.
Journal of insect physiology (2009-11-26)
Chiou Ling Chang, Thomas A Coudron, Cynthia Goodman, David Stanley, Shiheng An, Qisheng Song
ZUSAMMENFASSUNG
Culture medium supplemented with wheat germ oil (WGO) causes physiological reactions, such as increased fecundity and mobility, in some insects. Although the impact of WGO on insect physiology is important, the mechanisms of these actions are poorly understood. In this paper, we test the hypothesis that the addition of WGO to medium developed for larval oriental fruit flies modulates gene expression in the corresponding adults. We separately reared larvae of Bactrocera dorsalis on diets lacking or supplemented with WGO, and analyzed for expressed proteins in the resulting adult males and females by 2D-electrophoresis. Analysis of the gels revealed significant changes in expression levels of >70 proteins, 64 of which were identified by mass spectrometric analysis on MALDI-TOF/TOF. Apparent changes in expression levels for 6 of these proteins were confirmed by quantitative real-time PCR, showing that the changes in mRNA expression were reflected in changes in protein expression. These findings support the hypothesis that one mechanism of WGO actions in insect nutrition is the modulation of gene expression.