Merck
  • [The effect of warfarin was potentiated by an antitussive. Cocillana-Etyfin increased the PK(INR) value].
[The effect of warfarin was potentiated by an antitussive. Cocillana-Etyfin increased the PK(INR) value].
Lakartidningen (2009-12-17)
Henrik Lövborg, Ingela Jacobsson, Staffan Hägg