Merck
Evaluation of "Myrrh extract" against Schistosoma mansoni: a histological study.
Journal of the Egyptian Society of Parasitology (2010-05-28)
Ahmed M A Massoud, Faika H El Ebiary, Suzi H Ibrahim, Hanan A A Saleh, Hazem H M Khalil
ZUSAMMENFASSUNG
This study investigated the effect of myrrh extract on different developmental stages of Schistosoma mansoni. Sixty albino mice were used and divided into three main groups: GI (control group), GII (infected group) and GIII (infected-treated group). The last group was further divided into 3 subgroups where the drug was administered in a dose of 500 mg/kg body weight for 5 days starting on the 1st day PI for IIIA, on the 21st day PI for IIIB and on the 45th day PI for IIIC. A morphometric study was performed for the mean number and perimeter of granulomas. In GII, typical bilharzial granulomas were frequently encountered in the portal tracts with numerous eosinophils, collagen fiber deposition and reticular fiber condensation. Hepatocytes revealed vacuolation, nuclear affection and depletion of glycogen. In GIII, granulomas were less frequently observed with apparent decrease of eosinophils. The maximum effect of the drug was observed in SGs IIIB and IIIC as detected by significant decrease in the mean number and size of granulomas, paucity of eosinophils, decreased fibrosis and reticular fibers and the restoration of the glycogen content in the hepatocytes. The present data proved that myrrh has a valuable schistosomicidal effect against different stages of S. mansoni. This chemotherapeutic effect was more evident when the drug was given to infected mice on the 21st as well as on the 45th day PI.