Merck
[Determination of tomatine in Solanum cathayanum by RP-HPLC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-06-26)
Jianghong Guo, Yuan Zhou, Rong Liu, Mingxia Xie, Kun Zou
ZUSAMMENFASSUNG
To develop an HPLC method for determination of tomatine in the dried Solanum cathayanum of China Hubei Enshi. The analysis was performed on a YMC-Pack ODS-AA column (4. 6 mm x 150 mm, 5 microm) eluted with acetonitrile and water in gradient mode. The concentration of acetonitrile in the mobile phase changes from 20% to 100% within 60 minutes. The detection wavelength was set 203 nm. The flow rate was 1.0 mL x min(-1) and column temperature was set at 30 degrees C. The linear relationship of tomatine was determined within the range from 0.1-0.6 g x L(-1) (r = 0.999 7). The average recovery as 98.93% with RSD 1.2%. A convenient and reliable method was developed to determine the content of tomatine in the dried S. cathayanum.