Merck
  • Effect of oral exposure to fenitrothion and 3-methyl-4-nitrophenol on splenic cell populations and histopathological alterations in spleen in Wistar rats.
Effect of oral exposure to fenitrothion and 3-methyl-4-nitrophenol on splenic cell populations and histopathological alterations in spleen in Wistar rats.
Human & experimental toxicology (2010-07-14)
Qing Li, Maiko Kobayashi, Hirofumi Inagaki, Yukiyo Hirata, Shigeru Sato, Masamichi Ishizaki, Ai Okamura, Dong Wang, Tamie Nakajima, Michihiro Kamijima, Tomoyuki Kawada
ZUSAMMENFASSUNG
Fenitrothion (FNT) is used throughout the world as an insecticide in agriculture. To investigate the effect of FNT on the splenocytes and the underlying mechanism, FNT and its main metabolite, 3-methyl-4-nitrophenol (MNP), were administered orally to Wistar rats in daily doses of 0, 5 and 10 mg/kg, 4-5 days/week for 9 weeks. Splenocytes were harvested from control and exposed rats, and the following cell phenotypes were quantified by flow cytometry: (1) B cells (PE-CD45RA), (2) T cells (FITC-CD3), (3) T cell subsets (PE-CD4 and PerCPCD8), (4) natural killer (NK) cells (FITC-CD161a), (5) macrophages (FITC-CD11b), and (6) granulocyte (PE-granulocyte). Body weight, weight of the spleen, and histopathological alterations of spleens were also examined. The percentage of splenic CD8+ T cells and the ratio of CD8/CD4 in the group receiving 10 mg/kg FNT, and the percentages of splenic CD3+ and CD8+ T cells in the group receiving 10 mg/kg MNP were significantly decreased compared with those in the controls. FNT exposure also significantly decreased the weight of the spleen and body weight. In addition, apoptotic lymphocytes in spleen were observed in FNT-exposed rats under transmission electron microscope. However, FNT and MNP exposures did not affect splenic NK cells, B cells, macrophages, and granulocytes. The above findings indicate that FNT and MNP may selectively affect splenic T cells in rats.