Merck
  • Indoor use of plastic sheeting impregnated with carbamate combined with long-lasting insecticidal mosquito nets for the control of pyrethroid-resistant malaria vectors.
Indoor use of plastic sheeting impregnated with carbamate combined with long-lasting insecticidal mosquito nets for the control of pyrethroid-resistant malaria vectors.
The American journal of tropical medicine and hygiene (2010-08-05)
Armel Djènontin, Fabrice Chandre, K Roch Dabiré, Joseph Chabi, Raphael N'guessan, Thierry Baldet, Martin Akogbéto, Vincent Corbel
ZUSAMMENFASSUNG
The combined efficacy of a long-lasting insecticidal net (LLIN) and a carbamate-treated plastic sheeting (CTPS) or indoor residual spraying (IRS) for control of insecticide-resistant mosquitoes was evaluated in experimental huts in Burkina Faso. Anopheles gambiae from the area is resistant to pyrethroids and to a lesser extent, carbamates. Relatively low mortality rates were observed with the LLIN (44%), IRS (42%), and CTPS (52%), whereas both combinations killed significantly more mosquitoes (~70% for LLIN + CTPS and LLIN + IRS). Blood feeding by An. gambiae was uninhibited by IRS and CTPS compared with LLIN (43%), LLIN + CTPS (58%), and LLIN + IRS (56%). No evidence for selection of the kdr and ace-1(R) alleles was observed with the combinations, whereas a survival advantage of mosquitoes bearing the ace-1(R) mutation was observed with IRS and CTPS. The results suggest that the combination of the two interventions constitutes a potential tool for vector-resistance management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Bendiocarb, PESTANAL®, analytical standard