Merck
  • Determination of plasma catecholamines via condensation with diphenylethylenediamine: simplification of the procedure.
Determination of plasma catecholamines via condensation with diphenylethylenediamine: simplification of the procedure.
Journal of chromatography (1990-11-30)
P Husek, J Malíková, G Herzogová
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N′-Diphenylethylendiamin, 98%