Merck
  • Validation and clinical application of an LC-ESI-MS/MS method for simultaneous determination of tolmetin and MED5, the metabolites of amtolmetin guacil in human plasma.
Validation and clinical application of an LC-ESI-MS/MS method for simultaneous determination of tolmetin and MED5, the metabolites of amtolmetin guacil in human plasma.
Biomedical chromatography : BMC (2010-09-21)
Kishore Kumar Hotha, D Vijaya Bharathi, S Sirish Kumar, Y Narsimha Reddy, Pankaj Chatki, L K Ravindranath, K N Jaya Veera, Ramesh Mullangi
ZUSAMMENFASSUNG
A highly sensitive, rapid assay method has been developed and validated for the simultaneous estimation of tolmetin (TMT) and MED5 in human plasma with liquid chromatography coupled to tandem mass spectrometry with electrospray ionization in the positive-ion mode. A simple solid-phase extraction process was used to extract TMT and MED5 along with mycophenolic acid (internal standard, IS) from human plasma. Chromatographic separation was achieved with 0.2% formic acid-acetonitrile (25:75, v/v) at a flow rate of 0.50 mL/min on an X-Terra RP(18) column with a total run time of 2.5 min. The MS/MS ion transitions monitored were 258.1 → 119.0 for TMT, 315.1 → 119.0 for MED5 and 321.2 → 207.0 for IS. Method validation and clinical sample analysis were performed as per FDA guidelines and the results met the acceptance criteria. The lower limit of quantitation achieved was 20 ng/mL and the linearity was observed from 20 to 2000 ng/mL, for both the anlaytes. The intra-day and inter-day precisions were in the range 3.27-4.50 and 5.32-8.18%, respectively for TMT and 4.27-5.68 and 5.32-8.85%, respectively for MED5. This novel method has been applied to a clinical pharmacokinetic study.