Merck
  • Male mediated teratogenic potential evaluation of new antifertility compound SMA in rabbit (Oryctolagus cuniculus).
Male mediated teratogenic potential evaluation of new antifertility compound SMA in rabbit (Oryctolagus cuniculus).
Contraception (1990-08-01)
N Sethi, R K Srivastava, R K Singh
ZUSAMMENFASSUNG
The male mediated teratogenic potential of a new antifertility compound, styrene maleic anhydride (SMA), was evaluated in rabbits. No teratogenic potential was observed at the doses of 1.25 mg, 2.5 mg and 5.0 mg used in our experiment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(styrol-alt-Maleinsäure) Natriumsalz -Lösung, 13 wt. % in H2O