Merck
  • Small plasmids carrying vga(A) or vga(C) genes mediate resistance to lincosamides, pleuromutilins and streptogramin A antibiotics in methicillin-resistant Staphylococcus aureus ST398 from swine.
Small plasmids carrying vga(A) or vga(C) genes mediate resistance to lincosamides, pleuromutilins and streptogramin A antibiotics in methicillin-resistant Staphylococcus aureus ST398 from swine.
The Journal of antimicrobial chemotherapy (2010-09-30)
Kristina Kadlec, Constança F Pomba, Natacha Couto, Stefan Schwarz
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pleuromutilin, ≥95%