- A fatally mistaken fruit juice drink: an unordinary way of cocaine intoxication.
A fatally mistaken fruit juice drink: an unordinary way of cocaine intoxication.
Journal of forensic and legal medicine (2010-11-09)
R García-Repetto, M P Giménez, M C Martinez, M L Soria
PMID21056879
ZUSAMMENFASSUNG
Cocaine is one of the drugs of abuse more frequently consumed in Spain. Furthermore, Spain due to its geographical position is used by trafficker's organizations as the port of entrance of cocaine in the European Union. We present here a case of a fatal intoxication caused by a mistake in the cocaine distribution net in our country. Cocaine was concealed in a tropical juice only sold by the Internet.