Merck
  • Muscarinic receptors within the ventromedial hypothalamic nuclei modulate metabolic rate during physical exercise.
Muscarinic receptors within the ventromedial hypothalamic nuclei modulate metabolic rate during physical exercise.
Neuroscience letters (2010-11-26)
Samuel Penna Wanner, Juliana Bohnen Guimarães, Washington Pires, Umeko Marubayashi, Nilo Resende Viana Lima, Cândido Celso Coimbra
ZUSAMMENFASSUNG
The involvement of muscarinic cholinoceptors within the ventromedial hypothalamic nuclei (VMH) on the exercise-induced increase in oxygen consumption (VO(2)) was investigated. Rats were fitted with bilateral cannulae into the VMH for local delivery of drugs. On the day of the experiments, the animals were submitted to running exercise (20 m/min; 5% grade) until the point of fatigue. VO(2) was continuously measured after bilateral injections of either 0.2 μL of 5 × 10(-9)mol methylatropine or 0.15M NaCl solution into the VMH. Control experiments were conducted in freely moving rats on the treadmill. Muscarinic blockade within the VMH reduced time to fatigue by 32% and enhanced the increase in VO(2) from the 8th until the 17th min of exercise when compared to the control trial. In fact, time to fatigue was negatively correlated to the rate of increase in VO(2) (r(2)=0.747; P<0.001). However, bilateral injections of methylatropine in freely moving rats did not change VO(2) in comparison to saline injections. In conclusion, muscarinic cholinoceptors within the VMH are activated during exercise to modulate the increase in metabolic rate. Furthermore, blocking muscarinic transmission leads to a faster increase in VO(2) that is associated with the early interruption of exercise.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Methylatropinbromid, European Pharmacopoeia (EP) Reference Standard