Merck
  • Therapeutic efficacy of artesunate-amodiaquine combinations and the plasma and saliva concentrations of desethylamodiaquine in children with acute uncomplicated Plasmodium falciparum malaria.
Therapeutic efficacy of artesunate-amodiaquine combinations and the plasma and saliva concentrations of desethylamodiaquine in children with acute uncomplicated Plasmodium falciparum malaria.
American journal of therapeutics (2010-12-31)
Akintunde Sowunmi, Grace O Gbotosho, Christian T Happi, Titilope M Okuboyejo, Abayomi O Sijuade, Obaro S Michael, Elsie O Adewoye, Onikepe Folarin
ZUSAMMENFASSUNG
The treatment efficacy of artesunate-amodiaquine (AQ) coformulated or copackaged, and the plasma and saliva concentrations of desethylamodiaquine (DEAQ), the active metabolite of AQ, were evaluated in 120 and 7 children, respectively, with uncomplicated Plasmodium falciparum malaria treated with oral daily doses of the 2 formulations for 3 days. All children recovered clinically. Fever clearance (1.1 ± 0.2 vs 1.0 ± 0 days) and parasite clearance times (21.1 ± 10.2 vs 19.0 ± 7.0 hours) in artesunate-AQ coformulated and artesunate-AQ copackaged treated children, respectively, were similar. All children remained aparasitemic for at least 28 days. Blood and saliva samples were collected over 35 days and DEAQ in plasma and saliva was determined by high-performance liquid chromatography. DEAQ was detectable in plasma and saliva within 40 minutes of oral administration of artesunate-AQ. DEAQ concentrations 7 days after the start of therapy were 247.8 and 125.1 ng/mL in plasma and saliva, respectively. The concentration-time curves of plasma and saliva in declining phases were approximately parallel giving a similar half-life of 169.1 ± 16.4 and 142.8 ± 6.5 hours in plasma and saliva, respectively. Clearance from plasma and saliva was also similar (335.6 and 443.4 mL·h·kg, respectively). Area under concentration-time curves (AUC0-35d) for plasma and saliva were 94,744.9 and 74,004.2 ng·mL·h, respectively. In general, Saliva-plasma concentration ratio was 0.25-0.4. DEAQ concentrations in saliva may be useful for monitoring therapy and for the evaluation of the disposition of AQ in children with falciparum malaria treated with AQ-based combination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
N-Desethylamodiaquine dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®