Merck
  • [Effectiveness of duspatalin therapy in the treatment and prevention of post-cholecystectomy syndrome].
[Effectiveness of duspatalin therapy in the treatment and prevention of post-cholecystectomy syndrome].
Vestnik Rossiiskoi akademii meditsinskikh nauk (2011-03-15)
E M Lipnitskiĭ, O V Isakova, V A Movchun, M D Apdatskaia
ZUSAMMENFASSUNG
Results of evaluation of the efficiency of myotropic spasmolytic Duspatalin during long-term therapy and preventive treatment of functional post-cholecystectomy syndrome are presented. The influence of the treatment on manifestations of clinical symptoms, quality of a life estimated based on a visual-analog scale, and intestinal microbiocenosis (changes in the activity of short-chain fatty acids) are discussed.