- Ingestions of hydrocodone, carisoprodol, and alprazolam in combination reported to Texas poison centers.
Ingestions of hydrocodone, carisoprodol, and alprazolam in combination reported to Texas poison centers.
Journal of addictive diseases (2011-04-15)
Mathias B Forrester
PMID21491292
ZUSAMMENFASSUNG
The combination of hydrocodone, carisoprodol, and alprazolam is subject to abuse. Ingestions of this drug combination reported to Texas poison centers during 1998-2009 were identified (totaling 1,295 cases) and the distribution of ingestions by selected factors was determined. The number of cases increased from 0 in 1998 to 200 in 2007, and then decreased to 132 in 2009. The counties in eastern and southeastern Texas accounted for 80.9% of the cases. Of the patients, 57.3% were women and 94.6% were age 20 or older. Suspected attempted suicide accounted for 59.3% of the cases and intentional misuse or abuse for 27.3%.