Merck
  • Oxidation of 20-hydroxyleukotriene B4 to 20-carboxyleukotriene B4 by human neutrophil microsomes. Role of aldehyde dehydrogenase and leukotriene B4 omega-hydroxylase (cytochrome P-450LTB omega) in leukotriene B4 omega-oxidation.
Oxidation of 20-hydroxyleukotriene B4 to 20-carboxyleukotriene B4 by human neutrophil microsomes. Role of aldehyde dehydrogenase and leukotriene B4 omega-hydroxylase (cytochrome P-450LTB omega) in leukotriene B4 omega-oxidation.
The Journal of biological chemistry (1990-03-15)
H Sumimoto, S Minakami
ZUSAMMENFASSUNG
Leukotriene B4 (LTB4), a potent chemoattractant for leukocytes, is catabolized by human neutrophils via omega-oxidation. Neutrophil microsomes are known to oxidize 20-hydroxy-LTB4 (20-OH-LTB4) to its 20-oxo and 20-carboxy derivatives in the presence of NADPH. This activity has been ascribed to LTB4 omega-hydroxylase (cytochrome P-450LTB omega), a conclusion supported by our finding of the reversal of carbon monoxide inhibition by 450 nm light and by competitive inhibition studies. The oxidation of 20-oxo-LTB4 to 20-carboxy-LTB4 is also catalyzed by microsomes fortified with 1 mM NAD+, and this activity is not affected by cytochrome P-450LTB omega inhibitors. The evidence is compatible with involvement of a disulfiram-insensitive aldehyde dehydrogenase in this second oxidation pathway. Interaction of the two pathways is evidenced by facilitation of NADPH-dependent oxidation of 20-OH-LTB4 by the addition of NAD+. This synergism may be explained by removal of the aldehyde intermediate by the NAD(+)-dependent aldehyde dehydrogenase. Taken together with the finding that the NAD(+)-dependent activity is severalfold higher than the NADPH-dependent one, the dehydrogenase may be important in the oxidation of 20-OH-LTB4 to 20-carboxy-LTB4.