Merck
  • Immunoadsorption of IgG onto second antibody covalently attached to Amino-Dylark beads for radioimmunoassays.
Immunoadsorption of IgG onto second antibody covalently attached to Amino-Dylark beads for radioimmunoassays.
Clinical chemistry (1990-03-01)
S E Kakabakos, G P Evangelatos, D S Ithakissios
ZUSAMMENFASSUNG
We present a solid-phase immobilization method for radioligand assays, using an immunoadsorption coating procedure of anti-triiodothyronine rabbit IgG (anti-T3 IgG) onto second antibody (sheep anti-rabbit IgG) covalently bound to Amino-Dylark beads. The second antibody was in excess, compared with the first antibody, thus eliminating reproducibility problems between immunoadsorptions. Beads coated with second antibody can be used to immobilize a variety of antigen-specific first antibodies. The amount of anti-T3 antibody required for solid-phase T3 radioimmunoassay (RIA) was only 10% more, per assay tube, than that utilized in liquid-phase T3 RIA, in which polyethylene glycol solution was the separation reagent; characteristics of assay performance were comparable. The immobilization procedure requires high-titer antisera or antigen-specific IgG and seems advantageous because of the decrease in antibody requirements without significant modification of antibody functionality.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(styrol-alt-Maleinsäure) Natriumsalz -Lösung, 13 wt. % in H2O