- Draft genome sequence of Shewanella sp. strain HN-41, which produces arsenic-sulfide nanotubes.
Draft genome sequence of Shewanella sp. strain HN-41, which produces arsenic-sulfide nanotubes.
Journal of bacteriology (2011-08-27)
Dong-Hun Kim, Shenghua Jiang, Ji-Hoon Lee, Yong-Joon Cho, Jongsik Chun, Sang-Haeng Choi, Hong-Seog Park, Hor-Gil Hur
PMID21868804
ZUSAMMENFASSUNG
The dissimilatory metal reducing bacterium Shewanella sp. strain HN-41 was first reported to produce novel photoactive As-S nanotubes via reduction of As(V) and S(2)O(3)(2-) under anaerobic conditions. Here we report the draft genome sequence and annotation of strain HN-41.