Merck
  • A fluorescent probe for GM1 gangliosidosis related β-galactosidase: N-(dansylamino)hexylaminocarbonylpentyl-1,5-dideoxy-1,5-imino-D-galactitol.
A fluorescent probe for GM1 gangliosidosis related β-galactosidase: N-(dansylamino)hexylaminocarbonylpentyl-1,5-dideoxy-1,5-imino-D-galactitol.
Bioorganic & medicinal chemistry letters (2011-10-07)
Richard F G Fröhlich, Katrin Fantur, Richard H Furneaux, Eduard Paschke, Arnold E Stütz, Jacqueline Wicki, Stephen G Withers, Tanja M Wrodnigg
ZUSAMMENFASSUNG
N-(Dansylamino)hexylaminocarbonylpentyl-1,5-dideoxy-1,5-imino-D-galactitol, a strong competitive inhibitor of β-galactosidase, enhances residual β-galactosidase activities in fibroblasts and serves as lead en route to diagnostic compounds for tracking the fate of mutant β-gal as well as aberrant GM1 gangliosides by live cell imaging.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dulcit, ≥99% (GC)