Merck
  • Measurement of intracellular Na+ concentration by a Na+-sensitive fluorescent dye, sodium-binding benzofuran isophthalate, in porcine adrenal chromaffin cells--usage of palytoxin as a Na+ ionophore.
Measurement of intracellular Na+ concentration by a Na+-sensitive fluorescent dye, sodium-binding benzofuran isophthalate, in porcine adrenal chromaffin cells--usage of palytoxin as a Na+ ionophore.
Journal of neuroscience methods (1997-07-18)
S Ito, T Ohta, H Kadota, N Kitamura, Y Nakazato
ZUSAMMENFASSUNG
Palytoxin was found to equilibrate sodium ions (Na+) across the cell membrane much faster than dose gramicidin, which has been frequently used to calibrate the intracellular Na+ concentration ([Na+]in), in cells treated with a Na+-sensitive fluorescent dye, sodium-binding benzofuran isophthalate (SBFI). Palytoxin was capable of equilibrating Na+ in cells treated with SBFI-acetoxymethyl ester (SBFI-AM) and in voltage-clamped cells loaded with SBFI through a patch pipette. Nicotine caused a dose-dependent increase in ([Na+]in) in porcine adrenal chromaffin cells treated with SBFI-AM and caused a simultaneous increase in [Na+]in and inward current in the voltage-clamped cells loaded with SBFI. Palytoxin has an advantage of calibrating ([Na+]in) in a shorter time than dose gramicidin because of its powerful ionophoretic activity.