Merck
  • A sensitive capillary GC-MS method for analysis of topiramate from plasma obtained from single-dose studies.
A sensitive capillary GC-MS method for analysis of topiramate from plasma obtained from single-dose studies.
Biomedical chromatography : BMC (2012-01-10)
Jeannine M Conway, Angela K Birnbaum, Susan E Marino, James C Cloyd, Rory P Remmel
ZUSAMMENFASSUNG
Topiramate (Topamax®) is an antiepileptic medication used as adjunctive and monotherapy in patients with epilepsy and for migraine prophylaxis. A GC-MS assay was developed that was capable of detecting topiramate plasma concentrations following a single rectal or oral dose administration. Topiramate plasma samples were prepared by solid-phase extraction and were quantified by GC-MS analysis. The topiramate standard curves were split from 0.1 to 4 µg/mL and from 4 to 40 µg/mL in order to give a more accurate determination of the topiramate concentration. The accuracy of the standards ranged from 94.6 to 107.3% and the precision (%CV) ranged from 1.0 to 5.3% for both curves at all concentrations. The %CV for quality controls was <7.6%. The assay is both accurate and precise and will be used to complete future pharmacokinetic studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyheptamid