Merck
  • Voucher-based reinforcement for alcohol abstinence using the ethyl-glucuronide alcohol biomarker.
Voucher-based reinforcement for alcohol abstinence using the ethyl-glucuronide alcohol biomarker.
Journal of applied behavior analysis (2012-03-10)
Michael G McDonell, Donelle N Howell, Sterling McPherson, Jennifer M Cameron, Debra Srebnik, John M Roll, Richard K Ries
ZUSAMMENFASSUNG
This study assessed the effects of a contingency management (CM) intervention for alcohol consumption in 10 alcohol-dependent participants. An ABCA design was used. Vouchers were provided contingent on results of ethyl glucuronide (EtG) urine tests (an alcohol biomarker with a 2-day detection period) and alcohol breath tests during the C phase. The percentage of negative urines was 35% during the first baseline phase, 69% during the C phase, and 20% during the return-to-baseline phase. Results suggest that EtG urine tests may be a feasible method to deliver CM to promote alcohol abstinence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ethyl-β-D-glucuronid, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethyl-β-D-glucuronid, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®