Merck
Trials to address efficacy of midodrine 18 years after it gains FDA approval.
JAMA (2012-03-23)
Mike Mitka
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Midodrin -hydrochlorid, analytical standard, for drug analysis